Acid blue 161, black powder C.I. 15706

Synonyms: Disazo, Palatine Fast Blue BN, C.I.15706

0215259980 0215259925
IC15259925EA 38.52 USD
IC15259925 IC15259980
Acid blue 161, black powder C.I. 15706
Acid blue 161

Soluble in water.


Caution: Research Use Only (RUO).

Formula: C₄₀H₂₃CrN₄Na₂O₁₀S₂
MW: 881.738 g/mol
Melting Pt: 1900 °C
Storage Temperature: Ambient
MDL Number: MFCD00151053
CAS Number: 12392-64-2

Order Now

Specification Test Results

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR