Acid blue 29, black powder C.I. 20460

Synonyms: Disodium 4-amino-5-hydroxy-3-((3-nitrophenyl)azo)-6-(phenylazo)naphthalene-2,7-disulphonateMordant Blue 82C.I. 20460Disodium 4-amino-5-hydroxy-3-((3-nitrophenyl)azo)-6-(phenylazo)naphthalene-2,7-disulfonate

0215259625
IC15259625EA 41.67 USD
IC15259625
Acid blue 29, black powder C.I. 20460
Acid blue 29

Acid Blue 29 is an anionic disazo dye.


Slightly soluble in methyl cellosolve and water (20 mg/ml); very slightly soluble in ethanol (2 mg/ml).


Caution: Research Use Only (RUO).

Formula: C₂₂H₁₄N₆O₉S₂Na₂
MW: 616.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009960
CAS Number: 5850-35-1

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: λmax (water) 600 - 605 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR