Potassium hydrogen carbonate 99.7-100.5%, crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Potassium bicarbonate

0215255780
IC15255780EA 37.18 USD
IC15255780
Potassium hydrogen carbonate 99.7-100.5%, crystalline powder ACS
Potassium hydrogen carbonate

Potassium bicarbonate has been used in studies of renal disorders and the relationship of muscle injury to this process.


Potassium bicarbonate has been shown to inhibit the growth of Aspergillus parasiticus in Czapek's agar, and also aflatoxin synthesis.


Soluble in water (100 mg/mL); almost insoluble in alcohol.


Caution: Research Use Only (RUO).

Formula: KHCO₃
MW: 100.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6

Order Now

Specification Test Results

Identity Test Passes
Assay 99.7 - 100.5%
Insoluble Matter ≤0.01%
Chloride ≤0.001%
Phosphate ≤5 ppm
Sulfur Compounds ≤0.003%
Ammonium ≤5 ppm
Calcium (Ca) ≤0.002%
Magnesium (Mg) ≤0.001%
Heavy Metals ≤5 ppm
Iron ≤5 ppm
Sodium ≤0.03%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR