Lithium carbonate ≥99%, powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215253780
0215253790
IC15253780EA
67.06
USD
IC15253780
IC15253790
Lithium carbonate ≥99%, powder ACS
Lithium carbonate
Insoluble in Dilute HCl ≤0.01%; Chloride ≤0.005%; Nitrate (NO3) ≤5 ppm; Sulfur Compounds (as SO4) ≤0.2%; Iron (Fe) ≤0.002%; Calcium (Ca) ≤0.01%; Potassium (K) ≤0.01%; Sodium (Na) ≤0.1%
Formula:
Li₂CO₃ MW: 73.89 g/mol |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 |
Specification Test Results
Synonyms | Carbonic Acid Dilithium Salt; Carbonic Acid Lithium Salt; Dilthium Carbonate; Lithiumcarbonat; Carbolithium |
Form | White Powder |
Storage | Store at Room Temperature (15-30 °C). |
Assay | ≥99% |
Heavy Metals | ≤0.002% (as Pb) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...