Dodecyl-β-D-glucopyranoside, white powder

Warning

Synonyms: n-Dodecyl β-D-glucopyranosideDodecyl glucoside

0215248483 0215248480 0215248491
IC15248483EA 111.33 USD
IC15248483 IC15248480 IC15248491
Dodecyl-β-D-glucopyranoside, white powder
Dodecyl β-D-glucopyranoside

Dodecyl β-D-glucopyranoside is a long chain alkyl glycopyranoside and a classical nonionic amphiphile surfactant.


N-Dodecyl β-D-glucopyranoside is a classical nonionic amphiphile surfactant, which used in colloid research and micelle development. It may be used as a reference compound in long-chain alkyl glucoside separation and analysis procedures.


Non-ionic saccharide detergent used for the solubilization of membrane-bound proteins, such as G-Protein-Coupled Receptors, in native state


Soluble in methanol (200 mg/4 ml).

Formula: C₁₈H₃₆O₆
MW: 348.48 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063298
CAS Number: 59122-55-3

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Reference

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR