Acid red 52, powder C.I. 45100
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Kiton Red S
, Food Red No.106
, Lissamine rhodamine B-200
, Sulphorhodamine B sodium salt
, Sodium 4-(3,6-bis(diethylamino)xanthylium-9-yl)benzene-1,3-disulfonate
, Sodium 4-(3,6-bis(diethylamino)xanthylium-9-yl)benzene-1,3-disulphonate
, Sulforhodamine B sodium salt
Soluble in water (20 mg/mL) or ethanol (5 mg/mL). Creates a bluish-red solution with a yellow fluorescence.
Sulforhodamine B can be used as a protein tag - by sulfamido condensation with a proteinaceous substrate containing a primary amine function. Sulforhodamine B will fluoresce red. Because of its red fluorescence it can be used in conjuction with fluorescein isothiocyanate (which fluoresces yellowish-green).
Formula:
C₂₇H₃₀N₂O₇S₂ MW: 580.65 g/mol |
MDL Number:
MFCD00010180 CAS Number: 3520-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...