Sudan black B, dark powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Ceres black BN
, Solvent Black 3
, CI 26150
, Fat Black HB
0215208810
0215208825
IC15208825EA
86.13
USD
IC15208825
IC15208810
Sudan black B, dark powder
Sudan black B
Sudan Black B was used to detect native and oxidized low density lipoproteins (LDLs) after separation by capillary isotachophoresis (CITP).
- Dye Content: ~ 90%
It has been used for staining fat in animal tissues and in bacteria (Burdon's method), as well as the lipid portion of lipoprotein in polyacrylamide gel electrophoresis. It is a stain for chromosomes, Golgi apparatus, leucocyte (neutrophil) granules.
Soluble in ethanol; insoluble in water.
Formula:
C₂₉H₂₄N₆ MW: 456.55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006919 CAS Number: 4197-25-5 |
Specification Test Results
Identity Test | Passes |
UV/Vis: λmax (ethanol) | 598 ± 4 n |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...