ICNA02151813
0215181380 0215181305 0215181325
IC15181380EA 450.41 USD
IC15181380 IC15181305 IC15181325
ICNA02151813
1-Pentanesulfonic Acid Sodium Salt Monohydrate is used as an ion-associating reagent for HPLC analysis of peptides and proteins.
Sodium pentanesulfonate is utilized in the analysis of small organic molecular compounds, pharmaceutical products and metabolites, and peptides by such techniques as RP - HPLC and micellar electrokinetic chromatography.
Soluble in water (50 mg/mL).
Caution: Research Use Only (RUO).