ICNA02151790
0215179010
IC15179010EA 51.3 USD
IC15179010
ICNA02151790
Soluble in ammonium hydroxide (clear, colorless solution),Sodium hydroxide (50 mg/mL 1 M NaOH); slightly soluble in water (1.7 mg/mL).