γ-L-Glutamyl-p-Nitroanilide Hydrochloride. Powder
02151495.2 0215149505 0215149525 0215149501 0215149510 02151495.5
IC15149505EA 447.35 USD
IC15149505 IC151495.2 IC15149525 IC151495.5 IC15149501 IC15149510
γ-L-Glutamyl-p-Nitroanilide Hydrochloride. Powder
γ-L-Glutamyl-p-Nitroanilide hydrochloride is suitable for use as a substrate for g-Glutamyl transpeptidase.
Soluble in water (5 mg/mL - clear, light yellow solution) or 0.1 N HCl (1% w/v - clear, yellowish solution); soluble in methanol (100 mg/mL with slight heat and constant stirring).
Caution: Research Use Only (RUO).