Gum arabic, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Acacia gum
, Acacia
0215121890
0215121880
0215121891
IC15121880EA
54.38
USD
IC15121880
IC15121891
IC15121890
Gum arabic, Powder
Gum arabic
Total Ash ≤4.0%; Acid-insoluble Ash ≤0.5%; Lead ≤0.001%; Insoluble Residue ≤50.0 mg/5.0 g; Arsenic ≤3 ppm
MDL Number:
MFCD00081264 CAS Number: 9000-01-5 |
Specification Test Results
Synonyms | Gum Arabic; Arabic Gum; Australian Gum; Acacia Syrup; Indian Gum; Senegal Gum; Wattle Gum |
Form | Powder |
Storage | Store at room temperature (15-30 °C). |
Heavy Metals | ≤0.004% |
Moisture Content | ≤15% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...