Dihydromyrcenol, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215090625
IC15090625EA 93.11 USD
IC15090625
Dihydromyrcenol, liquid
Dihydromyrcenol
Formula: C₁₀H₂₀O
CAS Number: 18479-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR