2′-Deoxyadenosine-5′-monophosphate disodium hydrate ≥98%, white powder

Warning

Synonyms: 5'-Adenylic acid, 2'-deoxy-2′-Deoxyadenosine 5′-monophosphate sodium salt2-DeDisodium 2'-deoxy-5'-O-phosphonatoadenosineoxyadenylic acid

0215079501 02150795.1
IC15079501EA 420.85 USD
IC15079501 IC150795.1
2′-Deoxyadenosine-5′-monophosphate disodium hydrate ≥98%, white powder
2′-Deoxyadenosine-5′-monophosphate disodium hydrate

Soluble in water (50 mg/ml).


Caution: Research Use Only (RUO).

Formula: C₁₀H₁₂N₅O₆PNa₂
MW: 375.2 g/mol (anhy)
Melting Pt: 148 °C
Storage Temperature: Freezer
MDL Number: MFCD00065990
CAS Number: 151151-31-4

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
UV/VIS: λ max (water) 257 - 263, 203 - 209 nm
Moisture Recorded Result

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR