ICNA02150348
0215034880 0215034883 0215034891
IC15034880EA 97.77 USD
IC15034880 IC15034891 IC15034883
ICNA02150348
1-Aminocyclopropane-1-carboxylic acid is an intermediate in the conversion of methionine to ethylene during the ripening in apples. It also plays an important role in the biosynthesis of the plant hormone ethylene.
Soluble in water (49,00 - 51,00 mg/ml).