Fast blue B salt zinc double salt, powder C.I. 37235

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215020180 0215020110 0215020125
IC15020180EA 294.56 USD
IC15020180 IC15020125 IC15020110
Fast blue B salt zinc double salt, powder C.I. 37235
Fast Blue B salt zinc double salt

Fast blue B salt is useful for TLC of Aflatoxins.


Soluble in water( 4 mg/4 ml - clear solution).


Caution: Research Use Only (RUO).

Formula: C₁₄H₁₂Cl₄N₄O₂Zn
MDL Number: MFCD00075203
CAS Number: 14263-94-6
https://us.vwr.com/stibo/web/std.lang.all/45/62/36234562.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR