ICNA02150104
0215010401 0215010405 02150104.2
IC15010405EA 255.77 USD
IC15010405 IC150104.2 IC15010401
ICNA02150104
(Aminooxy)acetic Acid Hemihydrochloride undergoes condensation with aldehydes and ketones to form oximes. It inhibits aminobutyrate aminotransferase activity in vivo, thereby raising the level of gamma-aminobutyric acid in tissues.
Soluble in 100 mg/ml of water.
Caution: Research Use Only (RUO).