D-(+)-Xylose, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0210330091
0210330090
0210330080
IC10330080EA
53.17
USD
IC10330080
IC10330090
76102-588
D-(+)-Xylose, white powder
D(+)-Xylose
Xylose is the main building block for hemicellulose, the acid-catalysed degradation of hemicellulose gives furfural, a specialty solvent in industry and a precursor to synthetic polymers. In animal medicine, xylose is used to test for malabsorption by administration in water to the patient after fasting.
Soluble in water (clear,colorless solution), pyridine, and hot alcohol.
Caution: Research Use Only (RUO).
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Melting Pt: 155…157 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00151475 CAS Number: 58-86-6 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Optical Rotation | +18° ± 5° (c=4, H2O, NH4OH) |
Loss on Drying | ≤0.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...