Uric acid ∼99%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
1H-Purine-2,6,8(3H,7H,9H)-trione
0210321510
0210321580
0210321590
IC10321510EA
51.43
USD
IC10321510
IC10321580
IC10321590
Uric acid ∼99%, white powder
Uric acid
Soluble in alkaline solutions (1% in 1 N NaOH), glyurol; very slightly to insoluble in water; insoluble in ethanol, ether and alcohol.
Caution: Research Use Only (RUO).
Formula:
C₅H₄N₄O₃ MW: 168.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005712 CAS Number: 69-93-2 |
Specification Test Results
Purity | ~99% |
FTIR | Conforms to Standard |
Loss on Drying | ≤0,5% |
UV/VIS: λmax (water) | 230-235, 285-290 nm |
Residue on Ignition |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...