Sulfisoxazole, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210299325 0210299380
IC10299325EA 50.9 USD
IC10299325 IC10299380
Sulfisoxazole, crystalline powder
Sulfisoxazole

Soluble in Hydrochloric Acid (3 M); sparingly soluble in 96% V/V ethanol. Soluble in 10% hydrochloric acid (200 mg plus 6 mL-Clear colorless to faint yellow solution).

Formula: C₁₁H₁₃N₃O₃S
MW: 267.303 g/mol
MDL Number: MFCD00003150
CAS Number: 127-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR