ICNA02102976
0210297690 0210297680
IC10297690EA 316.93 USD
IC10297690 IC10297680
ICNA02102976
Succinylsulfathiazole is a sulfonamide. It is a competitive inhibitor of dihydropteroate synthetase. It is a dihydrofolate reductase (DHFR) inhibitor. used especially for treating gastrointestinal infections.
Succinylsulfathiazole is used to block the synthesis of folic acid. It is used to study folate deficiency. It suppress folate production by bacteria in the intestine hence used in prevention and treatment of gastrointestinal infections.
Slightly soluble in Water. Clear faint yellow solution at 50mg/mL in 1 M Sodium Hydroxide.
Succinylsulfathiazole is a competitive inhibitor of dihydropteroate synthetase. It is a dihydrofolate reductase (DHFR) inhibitor.