Semicarbazide hydrochloride ≥99%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Hydrazinecarboxamide hydrochloride

0210286490
IC10286490EA 143.15 USD
IC10286490
Semicarbazide hydrochloride ≥99%, powder
Semicarbazide hydrochloride

Soluble in Water (c = 10%).


Semicarbazide hydrochloride is used as urease substrate and MAO inhibitor. Derivatizing agent for carbonyl compounds as their semicarbazones which produces crystalline compounds with characteristic melting points. Also used in heterocyclic synthesis. Semicarbazone formation has been used to separate carbonyl compounds from mixtures by adsorption onto silica gel from a hydrocarbon solution. Regeneration is by hydrolysis with oxalic acid.

Formula: CH₆ClN₃O
MW: 111.53 g/mol
MDL Number: MFCD00013009
CAS Number: 563-41-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR