Pyridoxal hydrochloride, off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride

0210276705
IC10276705EA 76.75 USD
IC10276705
Pyridoxal hydrochloride, off-white powder
Pyridoxal hydrochloride

Pyridoxal hydrochloride is used for labeling amino acids and for their detection in the picomole range.


Finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.


Soluble in water (0.1% aq soln, 15 min., Clear, Colorless Solution) and in 95% alcohol.


Caution: Store Desiccated. Protect from light. Gradually darkens on exposure to air and sunlight

Formula: C₈H₁₀ClNO₃
MW: 203.62 g/mol
Melting Pt: 170…180 °C
Storage Temperature: Freezer
MDL Number: MFCD00012809
CAS Number: 65-22-5

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Solubility Test (0.1% aq soln, 15 min.) Clear, Colorless Solution C
pH (0.1% aq soln) 2.7 - 3.5
UV/VIS l max (water) 253-5, 290-1, 316-8 nm
Melting Point 170 - 180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR