(4-Nitrophenyl)-α-D-galactopyranoside, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

0210247101
IC10247101EA 201.78 USD
IC10247101
(4-Nitrophenyl)-α-D-galactopyranoside, Powder
(4-Nitrophenyl) α-D-galactopyranoside

p-Nitrophenyl-alpha-D-galactopyranoside is used as a substrate for α-galactosidase.


Soluble in water (5% w/v), HCl (50 mg/mL - clear, colorless solution) & methanol (100 mg/mL).

Formula: C₁₂H₁₅NO₈
MW: 301.26 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065050
CAS Number: 7493-95-0

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard C
Melting Point 164 - 176°C
Optical Rotation (c=1, 1N HCl) +240° ± 20°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR