Lysolecithin, Powder

Synonyms: [(2R)-3-acetyloxy-2-hydroxypropyl] 2-(trimethylazaniumyl)ethyl phosphate1-acetyl-sn-glycero-3-phosphocholineLysophosphatidyl Choline1,3-Lysolecithin

0210222390
IC10222390EA 1237.34 USD
IC10222390
Lysolecithin, Powder
Lysolecithin

Off-White Powder
Prepared enzymatically from lecithin.

Formula: C₁₀H₂₂NO₇P
MW: 299.26 g/mol
MDL Number: MFCD00131444
CAS Number: 9008-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR