Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl), Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Ethyl Glycinate Hydrochloride
, H-Gly-OEt.HCl
02101829.5
0210182901
IC10182901EA
207.35
USD
IC10182901
IC101829.5
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl), Powder
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl)
Glycine ethyl ester (GEE) is used in conjunction with N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide (EDC) for carboxyl-footprinting studies of proteins. The GEE/EDC protocol effects specific derivatization of glutamate and aspartate carboxyl side chains on intact proteins. This reaction is readily done under aqueous conditions at physiological pH.
Formula:
C₄H₁₀ClNO₂ MW: 139.58 g/mol |
MDL Number:
MFCD00012871 CAS Number: 623-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...