1,1'-Azobis(N,N-dimethylformamide), yellow crystals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: N1,N1,N2,N2-Tetramethyldiazene-1,2-dicarboxamideDiamide

0210152701 0210152705 02101527.1 02101527.5
IC101527.5EA 125.41 USD
IC101527.5 IC10152705 IC101527.1 IC10152701
1,1'-Azobis(N,N-dimethylformamide), yellow crystals
1,1'-Azobis(N,N-dimethylformamide)

Diamide is used as a thiol oxidizing agent. It has also been used to titrate protein glutathiolation to discriminate from other oxidative protein modifications. Diamide treatment increased protein glutathiolation in a concentration-dependent manner and had comparably little effect on protein-protein disulfide formation.


Soluble in water.


Caution: Research Use Only (RUO).

Order Now

Specification Test Results

Identity Test Passes
Melting Point 104 - 115 °C
UV/Vis: λmax (water) 295 - 300 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR