2'-Deoxyadenosine monohydrate ≥98%, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate
0210148105
0210148125
IC10148105EA
199.37
USD
IC10148105
IC10148125
2'-Deoxyadenosine monohydrate ≥98%, Powder
2'-Deoxyadenosine monohydrate
2′-Deoxyadenosine (2′-dAdo), a deoxyribonucleoside, may be used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.
Soluble in water (50 mg/ml).
Formula:
C₁₀H₁₃N₅O₃·1H₂O MW: 269.26 g/mol (251.24 anhy) |
MDL Number:
MFCD00149364 CAS Number: 16373-93-6 |
Specification Test Results
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
Melting Point | 186 - 196 °C |
UV/Vis: l max (water) | 259 ± 2 nm |
OD 250/OD 260 | 0.70 - 0.85 |
OD 280/OD 260 | 0.10 - 0.16 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...