Cholesterol hydrogen succinate, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Cholesteryl Hydrogen Succinate
, 5-Cholesten-3β-ol 3-hemisuccinate
, Succinic Acid Monocholesterol Ester
, Cholesteryl Hemisuccinate
, 3β-Hydroxy-5-cholestene 3-hemisuccinate
, CHS
0210137980
0210137905
0210137925
IC10137980EA
2019.15
USD
IC10137980
IC10137925
IC10137905
Cholesterol hydrogen succinate, Powder
Cholesterol hydrogen succinate
Appearance in solution: (5% in Toluene) clear, water white
Formula:
C₃₁H₅₀O₄ MW: 486.74 g/mol |
MDL Number:
MFCD00037705 CAS Number: 1510-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...