4-Chloro-7-nitrobenzofurazan ∼98%, Powder

Synonyms: NBD-ClNBD chloride4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole

0210136201
IC10136201EA 38.52 USD
IC10136201
4-Chloro-7-nitrobenzofurazan ∼98%, Powder
4-Chloro-7-nitrobenzofurazan

chloroform: soluble (50 mg/mL-clear to slightly hazy, faintly yellow to yellow)


Fluorogenic reagent for amino acids1. Also used to modify sulfhydryl groups and tyrosine residues in enzymes2. 4-Chloro-7-nitrobenzofurazan (NBD-chloride) was used in the synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine and functionalized hydroxynaphthofurazan. It was used in spectrophotometric and spectrofluorometric determination of clemastine hydrogen fumarate, loratadine, losartan potassium and ramipril in pharmaceutical formulations.

Formula: C₆H₂ClN₃O₃
MW: 199.55 g/mol
MDL Number: MFCD00005808
CAS Number: 10199-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR