Cesium acetate ≥99.0%, Powder, Optical Grade

Synonyms: Acetic acid cesium salt

0210131925
IC10131925EA 84.31 USD
IC10131925
Cesium acetate ≥99.0%, Powder, Optical Grade
Cesium acetate
Formula: H₃CCOOCs
MW: 191.95 g/mol
MDL Number: MFCD00013056
CAS Number: 3396-11-0

Order Now

Specification Test Results

Form White Powder
Storage Room Temperature, desiccate

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR