Uridine-5'-diphosphate trisodium salt ≥90%, white powder
Synonyms:
UDP sodium salt
0210120591
0210120525
0210120590
0210120580
IC10120525EA
25.35
USD
IC10120525
IC10120591
IC10120580
IC10120590
Uridine-5'-diphosphate trisodium salt ≥90%, white powder
Uridine-5'-diphosphate trisodium salt
Uridine-5'-diphosphate disodium salt is a nucleoside diphosphate.
- P2Y receptor agonist.
Uridine-5'-diphosphate Disodium Salt is a P2Y receptor agonist.
Soluble in water (5% w/v-clear).
Caution: Research Use Only (RUO).
Formula:
C₉H₁₄N₂Na₃O₁₂P₂ MW: 470.1 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00006524 CAS Number: 21931-53-3 |
Specification Test Results
Identity Test | Passes |
Purity | ≥90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...