L-α-Glycerophosphoryl Choline, Cadmium Chloride Complex (from egg yolk) ∼98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: sn-Glycero-3-phosphocholine 1:1 cadmium chloride adductα-GPC, Choline, Cadmium Chloride Complex

0210048701 02100487.5
IC10048701EA 345.55 USD
IC10048701 IC100487.5
L-α-Glycerophosphoryl Choline, Cadmium Chloride Complex (from egg yolk) ∼98%, white powder
L-α-Glycerophosphoryl Choline, Cadmium Chloride Complex
Formula: C₈H₂₀CdCl₂NO₆P
MW: 440.537 g/mol
Storage Temperature: Freezer
CAS Number: 64681-08-9
EINECS: 265-010-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR