APT-COM™ 3 Data Control System, Binder
Supplier: Binder
9053-0013 9053-0015 9053-0014
80079-152EA 2971.18 USD
80079-152 80079-150 80079-148
APT-COM™ 3 Data Control System, Binder
Software package APT-COM™ 3 Data control system for Windows to network up to 30 units or controllers with the communication interface RS 232, RS 422 or Ethernet for remote control, programming and data recording.
  • Individual user authorisation and identification through password protected access control or electronic signature
  • Complete and detailed audit trail for all system interventions
  • Data storage with protection against manipulation
  • Automatic data backup
  • Validability of overall system
APT-COM™ 3 is available at three different performance levels for use with Binder units equipped with a data interface: Full Version for applications according to FDA Directive 21 CFR part 11 in the GLP/GMP range, Standard Version with restricted data safety features and Basic Edition for simple data logging.