Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aso Adhesive Bandages, ASO

Supplier: ASO CORPORATION
CBD5293 CBD1321 CBD5118 CBD6418 CBD4025 CBD4024 CBD2022 CBD5B95 CBD4022 CBD5561 CBD1019 CBD8632 ACH2019 CBD4432 CBD5720 CBD5302 CBD2018 CBD4433 CBD4019 CBD4435 CBD2016 CBD2027 CBD4018
Careband adhesive bandages.



 

ORDER

Expand All / Collapse All
Description Size Material Dimensions Supplier No. VWR Catalog Number Unit Price Quantity
Butterfly Closure Bandages, Medium - - CBD6418 10028-218 Case of 12 Retrieving Restricted
Despicable Me Bandages - - 3/4 x 3" CBD5B95 10028-228 Case of 12 Retrieving Restricted
Sheer Bandages - - 2 x 4" CBD2016 10028-448 Case of 12 Retrieving Restricted
Sheer Spot Bandages - - 7/8" dia. CBD2022 10028-204 Case of 12 Retrieving Restricted
Sheer Mini Bandages - - 5/8" dia. CBD2027 10028-206 Case of 12 Retrieving Restricted
Fabric Bandages - - 1 x 3" CBD4019 10028-210 Case of 12 Retrieving Restricted
Fabric Bandages, Blue - - 1 x 3" CBD4433 10028-452 Case of 24 Retrieving Restricted
Fabric Fingertip Bandages - - CBD4025 10028-216 Case of 12 Retrieving Restricted
Fabric Fingertip Bandages, Blue - - CBD4432 10028-454 Case of 24 Retrieving Restricted
Fabric Knuckle Bandages - - CBD4024 10028-214 Case of 12 Retrieving Restricted
Fabric Knuckle Bandages, Blue - - CBD4435 10028-456 Case of 24 Retrieving Restricted
Fabric Spot Bandages - - 7/8" dia. CBD4022 10028-212 Case of 12 Retrieving Restricted
Fashion Strip Bandages, Assortment - - CBD5302 10028-230 Case of 12 Retrieving Restricted
Foam Bandages, Blue - - 1 x 3" CBD8632 10028-458 Case of 12 Retrieving Restricted
Garfield Strip Bandages - - 3/4 x 3" CBD5293 10028-220 Case of 12 Retrieving Restricted
Garfield Spot Bandages - - 7/8" dia. CBD5561 10028-222 Case of 12 Retrieving Restricted
Neon Strip Bandages - - CBD5118 10028-226 Case of 12 Retrieving Restricted
Plastic Bandages 25 x 76 mm (1 x 3") Plastic 1 x 3" CBD1019 89212-228 Case of 1200 Retrieving Restricted
Pack of 100 Retrieving Restricted
Waterproof Bandages - - 1 x 3" CBD1321 10028-464 Case of 12 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty