Cephalexin 50 mg/mL in ammonium hydroxide, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (6R,7R)-7-((R)-2-amino-2-phenylacetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

J67377-AC J67377-AD
AAJ67377-ADEA 94.28 USD
AAJ67377-AD AAJ67377-AC
Cephalexin 50 mg/mL in ammonium hydroxide, sterile filtered
Cephalexin
Formula: C₁₆H₁₉N₃O₅S
MW: 365.404 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00167148
CAS Number: 15686-71-2
EINECS: 239-773-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR