Ketoconazole 2 mg/ml in methanol, sterile filtered

Supplier: Alfa Aesar
Danger

Synonyms: (+/-)-cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine

J67352-XF
AAJ67352-XFEA 38.06 USD InStock
AAJ67352-XF
Ketoconazole 2 mg/ml in methanol, sterile filtered
Ketoconazole

Ketoconazole, 2 mg/ml in methanol, sterile-filtered

Formula: C₂₆H₂₈Cl₂N₄O₄
MW: 531.43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058579
CAS Number: 65277-42-1
EINECS: 265-667-4
UN: 1230
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR