Sulfamethoxazole 50 mg/ml in distilled water, sterile filtered

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Amino-N-(5-methylisoxazol-3-yl)benzenesulphonamide4-Amino-N-(5-methylisoxazol-3-yl)benzenesulfonamide

J67346-AC J67346-AD
AAJ67346-ADEA 111.57 USD
AAJ67346-AD AAJ67346-AC
Sulfamethoxazole 50 mg/ml in distilled water, sterile filtered
Sulfamethoxazole

Sulfamethoxazole, 50 mg/ml in distilled water, sterile-filtered

Formula: C₁₀H₁₁N₃O₃S
MW: 253.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010546
CAS Number: 723-46-6
EINECS: 211-963-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR