Miconazole nitrate 10 mg/ml in ethanol, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate

J67244-8EQ J67244-XF
AAJ67244-8EQEA 176.22 USD
AAJ67244-8EQ AAJ67244-XF
Miconazole nitrate 10 mg/ml in ethanol, sterile filtered
Miconazole nitrate
Formula: C₁₈H₁₄Cl₄N₂O·HNO₃
MW: 479.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00058161
CAS Number: 22832-87-7
EINECS: 245-256-6
UN: 1987
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR