Ethionamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Ethylpyridine-4-carbothioamide2-Ethylthioisonicotinamide

J66582-06
10067-200EA 203.73 USD
10067-200
Ethionamide 98%
Ethionamide
Formula: C₈H₁₀N₂S
MW: 166.24 g/mol
Boiling Pt: 297 °C
Melting Pt: 160…162 °C (dec.)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00057361
CAS Number: 536-33-4
EINECS: 208-628-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR