Fast green FCF ≥85% (dye content) for electrophoresis

Warning

Synonyms: Dihydrogen (ethyl)[4-[4-[ethyl(3-sulphonatobenzyl)amino](4-hydroxy-2-sulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene](3-sulphonatobenzyl)ammonium, disodium salt

J66570-14 J66570-06 J66570-22
AAJ66570-14EA 84.97 USD
AAJ66570-14 AAJ66570-06 AAJ66570-22
Fast green FCF ≥85% (dye content) for electrophoresis
Fast Green FCF
Formula: C₃₇H₃₄N₂Na₂O₁₀S₃
MW: 808.86 g/mol
Melting Pt: ca 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013053
CAS Number: 2353-45-9
EINECS: 219-091-5

Order Now

Specification Test Results

Appearance (Colour) Dark Brown
Form Powder
Assay (Unspecified), Dye Content Min. 85%
Solution Test 10% in NaOH, Blue Solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR