Cefotiam dihydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J66529-MC J66529-MF
10067-468EA 408.86 USD
10067-468 AAJ66529-MF
Cefotiam dihydrochloride
Cefotiam dihydrochloride

A broad spectrum antibiotic

Formula: C₁₈H₂₄ClN₉O₄S₃
MW: 562.079 g/mol
Boiling Pt: 94 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD09878605
CAS Number: 66309-69-1
EINECS: 266-312-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR