Minocycline hydrochloride 890-950 µg/mg
Supplier: Thermo Scientific Chemicals
|
Warning
|
J66429-03
J66429-ME
10067-462EA
237.36
USD
10067-462
10067-152
Minocycline hydrochloride 890-950 µg/mg
Minocycline hydrochloride
A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections
Minocycline hydrochloride, 890-950 µg/mg
Formula:
C₂₃H₂₇N₃O₇ Boiling Pt: 813 °C Melting Pt: 205…210 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00083669 CAS Number: 13614-98-7 EINECS: 237-099-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...