Flumequine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid
J66332-03
J66332-06
10067-128EA
160.26
USD
10067-128
10067-126
Flumequine 98%
Flumequine
Inhibits topoisomerases in Gram-negative bacteria.
Applications: Inhibits topoisomerases in gram negative bacteria.
Formula:
C₁₄H₁₂FNO₃ MW: 261.25 g/mol Melting Pt: 253…255 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00079298 CAS Number: 42835-25-6 EINECS: 255-962-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...