(R)-(+)-1-(1-Naphthyl)ethylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine(R)-1-(Naphthalen-1-yl)ethanamine

H66696-14 H66696-06
AAH66696-06EA 32.99 USD
AAH66696-06 AAH66696-14
(R)-(+)-1-(1-Naphthyl)ethylamine 97%
(R)-(+)-1-(1-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 152…153 °C/11 mm
Density: 1.060
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00064114
CAS Number: 3886-70-2
EINECS: 223-425-5
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR