1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride 98%

Warning

Synonyms: 1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride

H64862-06 H64862-14 H64862-22
AAH64862-22EA 590.52 USD
AAH64862-22 AAH64862-14 AAH64862-06
1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride 98%
1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride
Formula: C₁₅H₂₃NO₂
MW: 285.81 g/mol
Melting Pt: 174…178 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD06658143
CAS Number: 130198-05-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR