2-(Trifluoromethyl)thioxanthen-9-one 98%

Supplier: Thermo Scientific Chemicals
Warning

H64839-14 H64839-06 H64839-03
AAH64839-06EA 85.51 USD
AAH64839-06 AAH64839-14 AAH64839-03
2-(Trifluoromethyl)thioxanthen-9-one 98%
2-(Trifluoromethyl)thioxanthen-9-one
Formula: C₁₄H₇F₃OS
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00079785
CAS Number: 1693-28-3
EINECS: 216-893-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR