1,9-Dichlorononane 98%

Warning

H64561-22 H64561-06 H64561-14
AAH64561-06EA 34.13 USD
AAH64561-06 AAH64561-22 AAH64561-14
1,9-Dichlorononane 98%
1,9-Dichlorononane
Formula: C₉H₁₈Cl₂
MW: 197.15 g/mol
Boiling Pt: 258…262 °C
Density: 1.09
Flash Pt: >112 °C(234 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001024
CAS Number: 821-99-8
EINECS: 212-484-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR