4-Hexylphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Hexylphenylboronic Acid
H64555-03
H64555-MD
AAH64555-MDEA
53.09
USD
AAH64555-MD
AAH64555-03
4-Hexylphenylboronic acid 97%
4-Hexylphenylboronic acid
Formula:
C₁₂H₁₉BO₂ MW: 206.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039029 CAS Number: 105365-50-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...