2-(3-Chlorophenyl)benzimidazole 95%

Warning

H64245-06 H64245-MD H64245-03
AAH64245-03EA 254.19 USD
AAH64245-03 AAH64245-MD AAH64245-06
2-(3-Chlorophenyl)benzimidazole 95%
2-(3-Chlorophenyl)benzimidazole
Formula: C₁₃H₉ClN₂
Storage Temperature: Ambient
MDL Number: MFCD01438282
CAS Number: 22868-35-5

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR