4-Chloro-2,5-difluorobenzaldehyde 98%

H64112-03 H64112-MD H64112-06
AAH64112-03EA 178.71 USD
AAH64112-03 AAH64112-06 AAH64112-MD
4-Chloro-2,5-difluorobenzaldehyde 98%
4-Chloro-2,5-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782051
CAS Number: 879093-02-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR